


Xylene (mixture of isomers) ≥99.50%, Low-Metal (LM) grade semiconductor grade
Supplier: Transene
Certificates
About this item
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
Boiling Pt:140 °C
Melting Pt:–25 °C
Density:0.85 g/cc
Flash Pt:29 °C (closed cup)
UN:1307
ADR:3,II
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Xylene:99.5 min % Weight
- Color:10 APHA max.
- Acidity:0.3 ueq/g max.
- Residue after Evaporation:5000 ppb (umg/g) max.
- Moisture:0.02 % Weight max.
- Chloride:3000 ppb (umg/g) max.
- Phosphate:1000 ppb (umg/g) max.
- Sulfur Compounds:30000 ppb (umg/g) max.
- IR Spectra ACS:Pass
- Appearance:Clear Liquid
- Sub. Dark. BY H2SO4:Pass
- Density (at 25°C):To Report g/mL
- Aluminum:20 ppb (umg/g) max.
- Antimony:10 ppb (umg/g) max.
- Arsenic:10 ppb (umg/g) max.
- Barium:20 ppb (umg/g) max.
- Boron:20 ppb (umg/g) max.
- Cadmium:20 ppb (umg/g) max.
- Calcium:20 ppb (umg/g) max.
- Chromium:20 ppb (umg/g) max.
- Cobalt:20 ppb (umg/g) max.
- Copper:10 ppb (umg/g) max.
- Gallium:20 ppb (umg/g) max.
- Gold:20 ppb (umg/g) max.
- Iron:20 ppb (umg/g) max.
- Lead:20 ppb (umg/g) max.
- Lithium:20 ppb (umg/g) max.
- Magnesium:20 ppb (umg/g) max.
- Manganese:20 ppb (umg/g) max.
- Molybdenum:20 ppb (umg/g) max.
- Nickel:10 ppb (umg/g) max.
- Potassium:20 ppb (umg/g) max.
- Silicon:20 ppb (umg/g) max.
- Silver:20 ppb (umg/g) max.
- Sodium:20 ppb (umg/g) max.
- Tin:20 ppb (umg/g) max.
- Titanium:20 ppb (umg/g) max.
- Zinc:20 ppb (umg/g) max.
- Particle >1.0 μm:10 pcs/mL max.
- Particle >0.5 μm:150 pcs/mL max.