ContactUsLinkComponent
Disposable Heel Grounder, Transforming Technologies
Disposable Heel Grounder, Transforming Technologies
Catalog # 76489-732
Disposable Heel Grounder, Transforming Technologies
Catalog # 76489-732
Supplier Number:  HG4560L
Specifications
  • Color:
    Yellow
  • Description:
    Disposable heel grounder
  • Size:
    24" L
  • Cat. No.:
    76489-732
Specifications
About this item
Disposable heel grounders are comfortable to wear, easily identifiable, and use the entire volume of the fabric to provide protection.
  • Low cost method of grounding temporary personnel
  • High visibility yellow color
  • Disposable design is perfect for one time use
Constructed of a durable non-woven fabric impregnated with carbon, these disposable heel grounders use the entire volume of the fabric to provide protection.