Wolfgang Warmbier Two Point Probe, Transforming Technologies
76489-572
- Description:SRM mini probe, model 410, for resistance testers SRM110 and SRM200
- Cat. No.:76489-572
The Two-Point Probe is designed to measure surface resistance of small areas.
- Measures point-to-point surface resistance of small areas
- Spring-loaded contact pins
- Connects to meters with banana plugs
- Works with the SRM110 and SRM200 surface resistance meters
The probe works specifically with the SRM110 and SRM200 surface resistance meters, and may work with other models as well.