Polycarbonate Boxes, Crystal Industries
Supplier: CRYSTAL TECHNOLOGY BE
Certificates
About this item
Polycarbonate 81 or 100 cell 2" boxes hold most popular cryo and cold storage vials and fit into standard freezer racks.
- Boxes have clear lids with locator grids to facilitate and sample identification
- Polycarbonate boxes are designed to withstand temperature extremes of –196 to 121 °C
Polycarbonate boxes have drain holes for venting and safety and the keyed lid prevents misalignment of grid.