Order Entry
Puerto Rico
ContactUsLinkComponent
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥95%
Irritant
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥95%
Catalog # 76483-498
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Irritant
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥95%
Catalog # 76483-498
Supplier:  AAT BIOQUEST INC
Supplier Number:  22022
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    25 mg
  • Cat. No.:
    76483-498

Specifications

About this item

 :
 :557.467 g/mol
ADR:5,II
 :GHS07
CAS Number:150347-59-4
 :MFCD31812843
Formula:C₂₉H₁₉NO₁₁
MDL Number:MFCD00037466