About this item
High-pressure in-line filters and solvent mixers for longer column life and stable baseline.
- In-line filters available in 2.1 mm and 4.6 mm frit diameter
- Different porosities from 0.2 µm to 2 µm suit different dispersion needs
- Replaceable frits involve a one-time investment in the filter housing
- Available for Agilent and non-Agilent HPLC instruments
- Designed and manufactured to meet the highest quality requirements
- Jet-weaver mixer with optimized delay volume and highest mixing efficiency
Even small amounts of particulates can clog your column inlet, causing high column backpressure, retention-time shift, resolution loss, and shorter column life. Agilent offers high-pressure in-line filters designed to prevent blockages and extend column lifetime in any HPLC system, available in different dimensions and porosities. Agilent also provides a wide range of solvent mixers that deliver optimal mixing efficiency for exceptional baseline stability.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Related Information
Aglient Technologies Top Supplies to Optimize Your LC Workflow