Order Entry
Puerto Rico
ContactUsLinkComponent
ReadiUse™ Bacterial Cell Lysis Buffer *5X* (10 ml), AAT Bioquest
ReadiUse™ Bacterial Cell Lysis Buffer *5X* (10 ml), AAT Bioquest
Catalog # 76485-090
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ReadiUse™ Bacterial Cell Lysis Buffer *5X* (10 ml), AAT Bioquest
Catalog # 76485-090
Supplier:  AAT BIOQUEST INC
Supplier Number:  24100
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    5X concentrate
  • Description:
    ReadiUse™ bacterial cell lysis buffer
  • Size:
    10 ml
  • Cat. No.:
    76485-090

Specifications

About this item

Cell lysis refers to the breaking down of cells, and it is often used to analyze specific cellular compositions such as proteins, lipids, nucleic acids, reporter molecules, cell signal molecules and other small biomolecules.

  • ReadiUse™ reagents require minimal hands-on time