- Description:MycoLight™ Green JJ99 stain
- Size:100 µl
- Purity:95%
- Cat. No.:76483-904
MycoLight™ Green JJ99 stain is an excellent green-fluorescent nuclear and chromosome counterstain that is permeant to both prokaryotic and eukaryotic cell membranes.
- Cell-Permeable
- High DNA Affinity
- Bright Fluorescence