Order Entry
Puerto Rico
ContactUsLinkComponent
MycoLight™ Green JJ98 *5 mM in DMSO*, AAT Bioquest
MycoLight™ Green JJ98 *5 mM in DMSO*, AAT Bioquest
Catalog # 76483-902
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
MycoLight™ Green JJ98 *5 mM in DMSO*, AAT Bioquest
Catalog # 76483-902
Supplier:  AAT Bioquest
Supplier Number:  24000
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    MycoLight™ Green JJ98
  • Size:
    100 µl
  • Purity:
    95%
  • Cat. No.:
    76483-902

Specifications

About this item

MycoLight™ Green JJ98 is a green-fluorescent nuclear and chromosome stain that is permeant to both prokaryotic and eukaryotic cell membranes.

  • Cell-Permeable
  • High DNA Affinity
  • Bright Fluorescence
Caution: For Research Use Only