ContactUsLinkComponent
CytoFix™ Red Mitochondrial Stain, AAT Bioquest
  76485-166
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
CytoFix™ Red Mitochondrial Stain, AAT Bioquest
  76485-166
 :  23200
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    CytoFix™ Red mitochondrial stain
  • Purity:
    95%
  • Cat. No.:
    76485-166
  • No. of tests:
    500
CytoFix™ Red mitochondrial stain is a dye that selectively stains mitochondria independent of mitochondrial membrane potential gradient.
  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Useful for Multicolor Imaging
  • Robust
  • Retained After Fixation
 : For Research Use Only