Order Entry
Puerto Rico
ContactUsLinkComponent
CellPaint™ TSP membrane stain, AAT Bioquest
CellPaint™ TSP membrane stain, AAT Bioquest
Catalog # 76485-108
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
CellPaint™ TSP membrane stain, AAT Bioquest
Catalog # 76485-108
Supplier:  AAT Bioquest
Supplier Number:  22700
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    TSP
  • Size:
    10 mg
  • Purity:
    90%
  • Cat. No.:
    76485-108
  • No. of tests:
    500

Specifications

About this item

TSP is a styrylpyridine-based fluorescent membrane probe suitable for imaging plasma membranes in living cells and tissues.

  • Low Cytotoxicity
  • High Target Affinity
  • Bright Fluorescence
Caution: For Research Use Only