Order Entry
Puerto Rico
ContactUsLinkComponent
MitoLite™ Blue FX490, AAT Bioquest
MitoLite™ Blue FX490, AAT Bioquest
Catalog # 76483-694
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
MitoLite™ Blue FX490, AAT Bioquest
Catalog # 76483-694
Supplier:  AAT Bioquest
Supplier Number:  22674
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    MitoLite™ Blue
  • Purity:
    95%
  • Cat. No.:
    76483-694
  • No. of tests:
    500

Specifications

About this item

MitoLite™ Blue is a cationic dye that selectively accumulates in mitochondria probably via the mitochondrial membrane potential gradient.

  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Robust
Caution: For Research Use Only