Order Entry
Puerto Rico
ContactUsLinkComponent
LysoBrite™ Red DND-99, AAT Bioquest
LysoBrite™ Red DND-99, AAT Bioquest
Catalog # 76485-128
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
LysoBrite™ Red DND-99, AAT Bioquest
Catalog # 76485-128
Supplier:  AAT Bioquest
Supplier Number:  22647
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    LysoBrite™ Red DND-99
  • Purity:
    95%
  • Cat. No.:
    76485-128
  • No. of tests:
    500

Specifications

About this item

LysoBrite Red DND-99 is chemically same to the LysoTracker® Red DND-99 used for labeling and tracking acidic organelles in live cells (LysoTracker® is the trademark of ThermoFisher).

  • Lysosome-Specific Dye
  • Long-Term Use
  • Low Cytotoxicity
  • Stable After Fixation
Caution: For Research Use Only