ContactUsLinkComponent
Cell Meter™ RX780 Fixable Viability Dye, AAT Bioquest
Cell Meter™ RX780 Fixable Viability Dye, AAT Bioquest
Catalog # 76485-264
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Meter™ RX780 Fixable Viability Dye, AAT Bioquest
Catalog # 76485-264
Supplier:  AAT Bioquest
Supplier Number:  22536
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Cell Meter™ RX780 fixable cell stain
  • Cat. No.:
    76485-264
  • No. of tests:
    200
Specifications
About this item
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
  • Cell-Impermeable
  • Amine- And Thiol-Reactive
  • High Stability
  • Live/Dead Cell Labeling Dye
  • Fixable
Caution: For Research Use Only