- Description:Cell Meter™ BX590 fixable cell stain
- Cat. No.:76485-254
- No. of tests:200
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
- Cell-Impermeable
- Amine- And Thiol-Reactive
- High Stability
- Live/Dead Cell Labeling Dye
- Fixable