FunXite™-1, AAT Bioquest
FunXite™-1 is chemically same molecule to FUN® 1 (FUN® 1 is the trademark of ThermoFisher).
- Bicolor Fluorescence For Live/Dead Cell Differentiation
- Cell-Permeable
: For Research Use Only
- Description:FunXite™-1
- Size:100 µl
- Cat. No.:76485-076
- Purity:95%