Order Entry
Puerto Rico
ContactUsLinkComponent
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12], AAT Bioquest
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12], AAT Bioquest
Catalog # 76483-554
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12], AAT Bioquest
Catalog # 76483-554
Supplier:  AAT Bioquest
Supplier Number:  22120
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12]
  • Size:
    5 mg
  • Purity:
    95%
  • Cat. No.:
    76483-554

Specifications

About this item

The green fluorescent BODIPY FL C12 fatty acid can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.

  • Phospholipid Precursor
  • pH-Insensitive Fluorescence
  • Fluorescent In Aqueous and Lipids
Caution: For Research Use Only