Order Entry
Puerto Rico
ContactUsLinkComponent
 
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95%
Catalog # 76483-544
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95%
Catalog # 76483-544
Supplier:  AAT BIOQUEST INC
Supplier Number:  22076
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    25 mg
  • Cat. No.:
    76483-544

Specifications

About this item

 :
 :546.53 g/mol
CAS Number:53213-94-8
Formula:C₂₅H₂₇IN₂S₂
MDL Number:MFCD00075033