Order Entry
Puerto Rico
ContactUsLinkComponent
CytoTrace™ Green CMFDA, AAT Bioquest
CytoTrace™ Green CMFDA, AAT Bioquest
Catalog # 76483-488
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
CytoTrace™ Green CMFDA, AAT Bioquest
Catalog # 76483-488
Supplier:  AAT Bioquest
Supplier Number:  22017
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    CytoTrace™ Green CMFDA
  • Size:
    1 mg
  • Purity:
    95%
  • Cat. No.:
    76483-488

Specifications

About this item

CytoTrace™ Green CMFDA is an excellent replacement to CellTracker™ Green CMFDA (Invitrogen C2925, CellTracker™ is the trademark of Invitrogen).

  • Robust
  • High Multigenerational Cell Retention
  • Bright Signal
Caution: For Research Use Only