Order Entry
Puerto Rico
ContactUsLinkComponent
CytoTrace™ Red CFDA, AAT Bioquest
CytoTrace™ Red CFDA, AAT Bioquest
Catalog # 76483-486
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
CytoTrace™ Red CFDA, AAT Bioquest
Catalog # 76483-486
Supplier:  AAT Bioquest
Supplier Number:  22016
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    CytoTrace™ Red CFDA
  • Size:
    1 mg
  • Purity:
    90%
  • Cat. No.:
    76483-486

Specifications

About this item

CytoTrace™ Red fluorescent probe retains the spectral properties of Cy3/TRITC, thus its signal can be easily monitored using fluorescence microscopy or flow cytometry.

  • High Multigenerational Cell Retention
  • Bright Signal
Caution: For Research Use Only