Order Entry
Puerto Rico
ContactUsLinkComponent
Helixyte™ Gold Nucleic Acid Gel Stain *10000X DMSO Solution* (1 ml), AAT Bioquest
Helixyte™ Gold Nucleic Acid Gel Stain *10000X DMSO Solution* (1 ml), AAT Bioquest
Catalog # 76482-934
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Helixyte™ Gold Nucleic Acid Gel Stain *10000X DMSO Solution* (1 ml), AAT Bioquest
Catalog # 76482-934
Supplier:  AAT Bioquest
Supplier Number:  17595
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Helixyte™ Gold
  • Size:
    100 µg
  • Purity:
    95%
  • Cat. No.:
    76482-934

Specifications

About this item

Helixyte™ Gold is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Gold (SYBR® is the trademark of ThermoFisher).

  • Same Spectral Properties To SYBR® Gold
  • High Sensitivity
  • Useful For Nucleic Acid Labeling In Gels
Caution: For Research Use Only