Order Entry
Puerto Rico
ContactUsLinkComponent
Helixyte™ Green *10000X Aqueous PCR Solution*, AAT Bioquest
Helixyte™ Green *10000X Aqueous PCR Solution*, AAT Bioquest
Catalog # 76482-930
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Helixyte™ Green *10000X Aqueous PCR Solution*, AAT Bioquest
Catalog # 76482-930
Supplier:  AAT Bioquest
Supplier Number:  17592
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Helixyte™ Green
  • Size:
    1 ml
  • Purity:
    95%
  • Cat. No.:
    76482-930

Specifications

About this item

Helixyte™ Green is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Green (SYBR® is the trademark of ThermoFisher).

  • Same Spectral Properties To SYBR® Green
  • High Sensitivity
  • Useful For Nucleic Acid Labeling In Gels
Caution: For Research Use Only