Order Entry
Puerto Rico
ContactUsLinkComponent
DiTO™-3 [Equivalent To TOTO®-3], AAT Bioquest
DiTO™-3 [Equivalent To TOTO®-3], AAT Bioquest
Catalog # 76482-916
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
DiTO™-3 [Equivalent To TOTO®-3], AAT Bioquest
Catalog # 76482-916
Supplier:  AAT Bioquest
Supplier Number:  17576
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    DiTO™-3
  • Size:
    0.2 ml
  • Purity:
    95%
  • Cat. No.:
    76482-916

Specifications

About this item

DiTO™-3 is chemically equivalent to TOTO®-3 (TOTO® is the trademark of Invitrogen).

  • Cell-Impermeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence

5 mM DMSO Solution

Caution: For Research Use Only