Order Entry
Puerto Rico
ContactUsLinkComponent
CYTO-13, AAT Bioquest
CYTO-13, AAT Bioquest
Catalog # 76485-156
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
CYTO-13, AAT Bioquest
Catalog # 76485-156
Supplier:  AAT Bioquest
Supplier Number:  17553
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    CYTO-13
  • Size:
    5 ml
  • Purity:
    90%
  • Cat. No.:
    76485-156

Specifications

About this item

CYTO-13 is a fluorogenic, DNA-selective and cell-permeant dye for analyzing DNA content in living cells.

  • Cell-Permeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence

5 mM DMSO Solution

Caution: For Research Use Only