- Description:Nuclear Red™ LCS1
- Size:0.5 ml
- Purity:95%
- Cat. No.:76482-888
Our Nuclear Red™ LCS1 is a fluorogenic, DNA-selective and cell-permeant dye for analyzing DNA content in living cells.
- Cell-Permeable
- Useful For Multicolor Imaging
- Bright Fluorescence
5 mM DMSO Solution