Order Entry
Puerto Rico
ContactUsLinkComponent
Bodi Fluor™ R6G NHS Ester [equivalent to Bodipy® R6G, NHS Ester], AAT Bioquest
Bodi Fluor™ R6G NHS Ester [equivalent to Bodipy® R6G, NHS Ester], AAT Bioquest
Catalog # 76481-012
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Bodi Fluor™ R6G NHS Ester [equivalent to Bodipy® R6G, NHS Ester], AAT Bioquest
Catalog # 76481-012
Supplier:  AAT BIOQUEST INC
Supplier Number:  703
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Bodi Fluor™R6G
  • Size:
    5 mg
  • Purity:
    90%
  • Cat. No.:
    76481-012

Specifications

About this item

Bodi Fluor™R6G, NHS ester is the same molecule to Bodipy® R6G, NHS ester (Bodipy® is the trademark of Invitrogen).

  • Multiple Bioconjugation Applications
  • 90% Purity
Caution: For Research Use Only