Order Entry
Puerto Rico
ContactUsLinkComponent
Texas Red® Maleimide, Single Isomer, AAT Bioquest
Texas Red® Maleimide, Single Isomer, AAT Bioquest
Catalog # 76480-924
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Texas Red® Maleimide, Single Isomer, AAT Bioquest
Catalog # 76480-924
Supplier:  AAT Bioquest
Supplier Number:  483
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Texas Red® maleimide
  • Size:
    5 mg
  • Purity:
    95%
  • Cat. No.:
    76480-924

Specifications

About this item

Texas Red® maleimide is an excellent thiol-reactive fluorescent reagent for labeling thiol-containing biological molecules (such as peptides, proteins and thiol-modified oligonucleotides).

  • Identical spectra to Texas Red
  • High Stability
Caution: For Research Use Only