Order Entry
United States
ContactUsLinkComponent
LRB Red™ SE, AAT Bioquest
LRB Red™ SE, AAT Bioquest
Catalog # 76480-906
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
LRB Red™ SE, AAT Bioquest
Catalog # 76480-906
Supplier:  AAT BIOQUEST INC
Supplier Number:  471
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    LRB Red™
  • Size:
    10 mg
  • Environmentally Preferable:
  • Purity:
    90%
  • Cat. No.:
    76480-906

Specifications

About this item

Although Lissamine Rhodamine B chloride is one of the most popular labeling reagents of sulfonyl chloride, it is quite unstable in water. Even in anhydrous DMF, Lissamine Rhodamine B chloride tends to give a very complicated reaction mixture. It reacts with thiols, alcohols, phenols, aliphatic amines and aromatic amines indiscriminatingly. LRB Red™ is a succinimidyl ester and has the spectral properties identical to those of Lissamine Rhodamine B chloride. LRB Red™ is a superior replacement for Lissamine Rhodamine B chloride. LRB Red™ only reacts with aliphatic amines such as amino acids, peptides and proteins to give bright red fluorescent conjugates that are extremely stable. Compared to Lissamine Rhodamine B chloride, LRB Red™ has much higher labeling efficiency, and more importantly the resulted conjugates are much easier to be purified due to its much cleaner reactions. We strongly recommend that you consider using LRB Red™ to replace Lissamine Rhodamine B chloride for labeling peptides and oligonucleotides.

  • Identical spectra to Lissamine Rhodamine B chloride
  • High Labeling Efficiency
Caution: For Research Use Only