Order Entry
Puerto Rico
ContactUsLinkComponent
Cyanine 5 bissuccinimidyl ester ≥90% [equivalent to Cy5® bisNHS ester]
Cyanine 5 bissuccinimidyl ester ≥90% [equivalent to Cy5® bisNHS ester]
  76480-664
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
Cyanine 5 bissuccinimidyl ester ≥90% [equivalent to Cy5® bisNHS ester]
  76480-664
 :  157
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Size:
    1 mg
  • Cat. No.:
    76480-664

 

 

 :
 :1038.23 g/mol