Order Entry
Puerto Rico
ContactUsLinkComponent
 
p-GP/MDR1 Drug Interaction Assay Kit
Catalog # 103881-814
CAS Number:  
undefined
p-GP/MDR1 Drug Interaction Assay Kit
Catalog # 103881-814
Supplier Number:  HPGP-48
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    P-Glycoprotein / MDR1 Drug Interaction Assay Kit
  • Cat. No.:
    103881-814
  • Format:
    2× 48 assays in 96-well format
  • Species reactivity:
    Human

Specifications

About this item

Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from INDIGO’s Human P-Glycoprotein/MDR1 Drug Interaction Assay Kit, All-inclusive Cell-Based Luciferase Reporter Assay Kit, 2×48 format.

  • Rapidly assess drug candidates as either inhibitors, substrates, or non-substrates of P-glycoprotein
  • Kit includes kit contains two aliquots of HCT-Pgp cells, Cell Recovery Medium (CRM-p), Compound Screening Medium supplemented with daunorubicin (CSM+DR), Wash Buffer, two aliquots of Lysis Reagent, reference inhibitor Tariquidar, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)

The primary application of INDIGO Biosciences MDR1 drug interaction assay kit is to rapidly assess drug candidates as either inhibitors, substrates, or non-substrates of P-glycoprotein. This is accomplished by co-treating the cells with varying concentrations of the test drug and a fixed concentration of daunorubicin, a well-characterized fluorescent substrate for P-glycoprotein mediated transport. HCT-Pgp cells are a proprietary cell line that have been extensively selected for high-level expression of native P-glycoprotein. The kit contains two aliquots of HCT-Pgp cells and two black, sterile, collagen-coated 96-well assay plates. In addition to HCT-Pgp cells and assay plates, this kit includes Cell Recovery Medium (CRM-p) used to perform a rapid thaw of the –80 °C HCT-Pgp cells, Compound Screening Medium supplemented with daunorubicin (CSM+DR) used to prepare drug treatment media, Wash Buffer, two aliquots of Lysis Reagent, and the potent reference inhibitor Tariquidar.

Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C.