To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Retinoid X receptor panel (RXRa, RXRb, and RXRg) reporter assay kit
- Cat. No.:103881-548
- Format:3× 32 assays in 96-well format
- Species reactivity:Human
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Human RXR Reporter Assays Panel All-inclusive Cell-Based Luciferase Reporter Assay Kit, Including RXRa, RXRb, RXRg, each in 1×32 96-well format.
- Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
- Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed protocol
- Exceptional cell viability post-thaw
- Consistent results lot to lot (product quality data available upon request)
INDIGO's Retinoid X Receptors Panel (RXR alpha, RXR beta, and RXR gamma) assay kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to reporter cells for each RXR, optimized cell culture medium, a medium for diluting test compounds, a control agonist for each RXR, stable-glow luciferase detection reagent, detailed protocol, Protocol Quick Guide, and a cell culture-ready assay plate in strip-well format so that (if preferred) RXR alpha, RXR beta, and RXR gamma assays may be performed at different times.
This panel contains sufficient materials to perform 32 RXR alpha assays, 32 RXR beta assays, and 32 RXR gamma assays in 96-well plate format.