Order Entry
Puerto Rico
ContactUsLinkComponent
Human LXR Panel Luciferase Reporter Assay Kit
Human LXR Panel Luciferase Reporter Assay Kit
Catalog # 103881-510
CAS Number:  
undefined
Human LXR Panel Luciferase Reporter Assay Kit
Catalog # 103881-510
Supplier Number:  IB00321-48P
CAS Number:  

Specifications

  • Description:
    Liver X receptor panel (LXRa and LXRb) reporter assay kit
  • Cat. No.:
    103881-510
  • Format:
    2× 48 assays in 96-well format
  • Species reactivity:
    Human

Specifications

About this item

Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Human LXR Reporter Assays Panel All-inclusive Cell-Based Luciferase Reporter Assay Kit includes reporter cells for each LXR, optimized growth media, media for diluting test compounds, a control agonist for each LXR, luciferase detection reagent, a cell culture-ready assay plate in strip-well format.

  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)

INDIGO’s Human Liver X Receptor Panel (LXR alpha, LXR beta) assay kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to reporter cells for each LXR, optimized growth media, media for diluting test compounds, a control agonist for each LXR, luciferase detection reagent, a cell culture-ready assay plate in strip-well format so that (if preferred) LXR alpha and LXR beta assays may be performed at different times, detailed protocol, and a protocol quick guide.

This panel contains sufficient materials to perform 48 LXR alpha assays and 48 LXR beta assays, all in a single 96-well plate format.

Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.