USP and EP Vis Standards Set for Spectrophotometers, Thermo Scientific
Catalog # 76472-754
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:USP and EP Vis Standards Set, calibrated for use
with instruments with SBW of < 6.5 nm, 240-640 nm - Cat. No.:76472-754
Specifications
About this item
Reference Materials to quality performance verification (PV) tests in UV-Vis region. A set of filter standards for pharmacopeia compliance support.
- Wavelength accuracy
- Photometric accuracy
Wavelength Accuracy: Didymium glass filter
Photometric Accuracy: 4 neutral density filters calibrated for absorbance at 440, 465, 546.1, 590 and 635 nm. Absorbance values approximately 0.5, 1.0, 1.5 and 2.0.