Orders LinkContactUsLinkComponent
Paraffin Tapper
Paraffin Tapper
Supplier:   Mopec
Paraffin Tapper
Supplier:  Mopec
Certificates
Paraffin Tapper
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Designed to flatten tissue during the histology embedding process.
  • Stainless steel construction for durability and corrosion resistance
  • Ergonomic handle for comfortable, precise operation
  • Easy to clean and maintain for long-term use
  • Essential tool for pathology and histology workflows
Constructed from durable, laboratory-grade stainless steel, the Mopec Paraffin Tapper offers long-lasting performance and resistance to corrosion. Its ergonomic handle ensures a comfortable, controlled grip, reducing hand fatigue during repetitive use. The balanced design provides excellent control, making it ideal for use in histology, pathology, and research laboratories.
Product Family Options
Product Information
  • Availability
    Price
  • 76498-582
    Stainless steel paraffin tapper, 5×5×1.5 mm
    Stainless steel paraffin tapper, 5×5×1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $127.57
    $127.57
    Each
       $127.57
    Supplier Number: BG029
  • 76498-584
    Stainless steel paraffin tapper, 13×12×1.5 mm
    Stainless steel paraffin tapper, 13×12×1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $74.53
    $74.53
    Each
       $74.53
    Supplier Number: BG030
  • 76498-586
    Stainless steel paraffin tapper, 29×23×1.5 mm
    Stainless steel paraffin tapper, 29×23×1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $80.43
    $80.43
    Each
       $80.43
    Supplier Number: BG032
  • 76498-588
    Stainless steel paraffin tapper, 68×46×1.5 mm
    Stainless steel paraffin tapper, 68×46×1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $94.18
    $94.18
    Each
       $94.18
    Supplier Number: BG034
  • 76549-700
    Stainless steel paraffin tapper, 22×22×1.5 mm
    Stainless steel paraffin tapper, 22×22×1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $63.06
    $63.06
    Each
       $63.06
    Supplier Number: BG031
  • 76549-702
    Stainless steel paraffin tapper, 35×23×1.5 mm
    Stainless steel paraffin tapper, 35×23×1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $67.80
    $67.80
    Each
       $67.80
    Supplier Number: BG033