ContactUsLinkComponent
Coplin Staining Jar, Wide Top with Cover, Mortech®
Coplin Staining Jar, Wide Top with Cover, Mortech®
  76498-156
undefined
Coplin Staining Jar, Wide Top with Cover, Mortech®
  76498-156
 :  BA004
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Wide top coplin staining jar with cover
  • Cat. No.:
    76498-156
Heavy, clear glass staining jar with wide mouth and cover.
  • Glass construction
  • Eight molded grooves in glass hold up to sixteen 3×1" slides
Jar has eight slots, with each slot capable of holding two microscope slides. The wide top is convenient for transferring slides.