Order Entry
Puerto Rico
ContactUsLinkComponent
Human PPAR Panel Luciferase Reporter Assay Kit
Human PPAR Panel Luciferase Reporter Assay Kit
Catalog # 103881-492
CAS Number:  
undefined
Human PPAR Panel Luciferase Reporter Assay Kit
Catalog # 103881-492
Supplier Number:  IB00131-32P
CAS Number:  

Specifications

  • Description:
    PPAR panel (PPARa, PPARd and PPARg) reporter assay kit
  • Cat. No.:
    103881-492
  • Format:
    3× 32 assays in 96-well format
  • Species reactivity:
    Human

Specifications

About this item

Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. Human PPAR Reporter Assays Panel reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert.

  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)

INDIGO's Human PPAR Panel (PPAR alpha, PPAR delta, and PPAR gamma) assay kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to reporter cells for each PPAR, optimized growth media, media for diluting test compounds, a control agonist for each PPAR, luciferase detection reagent, detailed protocol, and a protocol quick guide, and a cell culture-ready assay plate in strip-well format so that (if preferred) PPAR alpha, PPAR delta, and PPAR gamma assays may be performed at different times.

This panel contains sufficient materials to perform 32 PPAR alpha assays, 32 PPAR gamma assays, and 32 PPAR delta assays, all in a single 96-well plate format.

Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.