ContactUsLinkComponent
5000-Place Slide Storage Cabinet, Mortech®
5000-Place Slide Storage Cabinet, Mortech®
Catalog # 76498-598
Supplier:  Mopec
undefined
5000-Place Slide Storage Cabinet, Mortech®
Catalog # 76498-598
Supplier:  Mopec
Supplier Number:  BH011
Specifications
  • Description:
    5000-place slide storage cabinet
  • Dimensions:
    15.75×18.75×5"
  • Cat. No.:
    76498-598
Specifications
About this item
Stackable, high-quality metal cabinets can be interlocked for stacking.
  • Heavy-duty metal construction
  • Baked enamel finish
  • Holds 5000 slides
  • Six (6) drawer cabinet
Each drawer includes a tab for labeling.