Orders LinkContactUsLinkComponent
Autopsy Head Drape and Framework
Autopsy Head Drape and Framework
Supplier:   Mopec
Autopsy Head Drape and Framework
Supplier:  Mopec
Certificates
Autopsy Head Drape and Framework
Autopsy Head Drape and Framework
1 Options Available Below
Product Details & Documents
Helps contain infectious and contaminated materials during autopsy procedures. Easily assembled with a wide variety of head, torso and/or appendage dissections.
  • Autopsy head drape kit includes framework and four polyethylene head drapes (0.03 mm thick, non-autoclavable)
  • Easily assembled with a wide variety of head, torso and/or appendage dissections
Product Family Options
Product Information
  • Material
    Dimensions
    Availability
    Price
  • 76498-428
    10 head drapes, without framework
    10 head drapes, without framework
    Discontinued
    Supplier Number: -
  • 76498-426
    Autopsy head drape and framework
    Autopsy head drape and framework
    Polyethylene
    26×20×4.5"
    Log in for availability
    Log in for availability
    Each
    Each - $579.71
    $579.71
    Each
       $579.71
    Supplier Number: BE010