Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trifluoroacetic acid
Trifluoroacetic acid neat Mobile Phase Modifier for HPLC/LCMS
Trifluoroacetic acid neat Mobile Phase Modifier for HPLC/LCMS
Trifluoroacetic acid neat Mobile Phase Modifier for HPLC/LCMS
SXMPK-TFAA-5PK
:
SPEX CERTIPREP LLC
:
Trifluoroacetic acid neat Mobile Phase Modifier for HPLC/LCMS
SXMPK-TFAA-5PK
:
SPEX CERTIPREP LLC
:
MPK-TFAA-5PK
:
Pack type:
Ampoule
Size:
5× 1 ml
Cat. No.:
SXMPK-TFAA-5PK
:
:
114.02 g/mol
:
Refrigerator
:
2699
:
8,I
:
GHS06
:
GHS05
:
GHS07
:
76-05-1
:
C₂HF₃O₂
:
MFCD00004169