Warmbier Metriso® B530 ESD Audit Kit, Transforming Technologies
Catalog # 76474-614
Supplier: Transforming Technologies
Specifications
- Description:Metriso® B530 ESD audit kit
- Cat. No.:76474-614
Specifications
About this item
Kit includes the relevant test instruments and equipment needed to perform ESD audits.
- Measure electrostatic fields
- Measure surface resistance
- Analyze ionizers performance
- Evaluate packaging materials
- Evaluate garments
- Measure flooring, work surfaces and furniture resistance
- Measure temperature and relative humidity
The Metriso® B530 ESD Audit Kit includes the relevant test instruments and equipment needed to perform ESD audits according to ANSI ESD.2020 and IEC 61340-5-1 and for qualification/verification of ESD control elements. The kit is built around the Metriso® B530 digital high resistance meter. It can take resistance measurements of 1 to 199 kΩ.