GemCell™ human serum AB is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material is defibrinated from source plasma AB. All donor units are tested for viral markers and found to be non-reactive.
- 0.1 µm sterile-filtered
- Gamma irradiation available
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76683-624
- 76683-636
- Origin:
- US
- US
- Storage Temperature:
- Store frozen at -20 to -10 °C
- Store frozen at -20 to -10 °C