Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Arg-Gly-Asp-Ser

Supplier:   Ambeed
Arg-Gly-Asp-Ser
Supplier:  Ambeed
Certificates
Arg-Gly-Asp-Ser

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Certifications: for Research Use Only

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 76687-302
  • 76687-306
  • 76687-300
  • 76687-304
  • Protein/Peptide Name:
  • Arg-Gly-Asp-Ser
  • Arg-Gly-Asp-Ser
  • Arg-Gly-Asp-Ser
  • Arg-Gly-Asp-Ser
  • CAS No.:
  • 91037-65-9
  • 91037-65-9
  • 91037-65-9
  • 91037-65-9
  • MDL No.:
  • MFCD00076452
  • MFCD00076452
  • MFCD00076452
  • MFCD00076452
  • Purity:
  • 98%
  • 98%
  • 98%
  • 98%
  • Molecular Weight:
  • 433.4170
  • 433.4170
  • 433.4170
  • 433.4170
  • Molecular Formula:
  • C₁₅H₂₇N₇O₈
  • C₁₅H₂₇N₇O₈
  • C₁₅H₂₇N₇O₈
  • C₁₅H₂₇N₇O₈

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $22.00
    $22.00
    Each (1mg)
       $22.00

    Supplier Number: A107716-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76687-302
    Protein/Peptide Name
    Arg-Gly-Asp-Ser
    CAS No.
    91037-65-9
    MDL No.
    MFCD00076452
    Purity
    98%
    Molecular Weight
    433.4170
    Molecular Formula
    C₁₅H₂₇N₇O₈
  • Glass bottle
    5 mg
    Log in for availability
    Log in for availability
    Each (5mg)
    Each (5mg) - $28.00
    $28.00
    Each (5mg)
       $28.00

    Supplier Number: A107716-5MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76687-306
    Protein/Peptide Name
    Arg-Gly-Asp-Ser
    CAS No.
    91037-65-9
    MDL No.
    MFCD00076452
    Purity
    98%
    Molecular Weight
    433.4170
    Molecular Formula
    C₁₅H₂₇N₇O₈
  • Glass bottle
    10 mg
    Log in for availability
    Log in for availability
    Each (10mg)
    Each (10mg) - $44.00
    $44.00
    Each (10mg)
       $44.00

    Supplier Number: A107716-10MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76687-300
    Protein/Peptide Name
    Arg-Gly-Asp-Ser
    CAS No.
    91037-65-9
    MDL No.
    MFCD00076452
    Purity
    98%
    Molecular Weight
    433.4170
    Molecular Formula
    C₁₅H₂₇N₇O₈
  • Glass bottle
    50 mg
    Log in for availability
    Log in for availability
    Each (50mg)
    Each (50mg) - $142.00
    $142.00
    Each (50mg)
       $142.00

    Supplier Number: A107716-50MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76687-304
    Protein/Peptide Name
    Arg-Gly-Asp-Ser
    CAS No.
    91037-65-9
    MDL No.
    MFCD00076452
    Purity
    98%
    Molecular Weight
    433.4170
    Molecular Formula
    C₁₅H₂₇N₇O₈

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.