Sulfo-SMCC Activated SureLight™ Peridinin-Chlorophyll Protein Complex (PerCP)
Catalog # 10159-606
Supplier: Columbia Biosciences
CAS Number:
Specifications
- Species:Human
- Size:1 mg
- Storage Conditions:Store at 4 °C
- Protein/Peptide Name:Activated Percp
- Tested Applications:Flow Cytometry, Cell-Based Assays, Microarrays and Microplate Applications
- Cat. No.:10159-606
- Supplier no.:D11-010
Specifications
About this item
Peridinin is a light-harvesting pigment associated with chlorophyll and found in the peridinin chlorophyll binding protein (PerCP); a light harvesting protein found in dinoflagellates. Commonly used in immunoassays such as FACS and flow cytometry.
- Emission: 676 nm
- Excitation: 483 nm