About this item
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The Human CD3E & CD3D Heterodimer-coupled Magnetic Beads is produced by coupling biotinylated Human CD3E & CD3D protein to streptavidin-conjugated magnetic beads. The pre-coupled beads are ready to use for capturing anti-Human CD3E & CD3D antibody from your sample with high specificity. The uniform size and large surface-to-volume ratio of the beads ensure highly efficent immunocapture in a simple, fast and convenient workflow with high reproducibilty of data.
Application Method:
a) Reconstitute the Beads following the COA. Wash and re-suspended the beads to a certain concentration by adding your dilution buffer.
b) Add the prepared beads to your samples.
c) Beads can be separated from your samples afterwards using a magnetic plate.
Specifications
- Catalog No:
- 103871-348
- 103871-346
- Storage Conditions:
- –20 °C, avoid light
- –20 °C, avoid light