Order Entry
Puerto Rico
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid Protein Titer Assay Kit, Acro Biosystems
SARS-CoV-2 Nucleocapsid Protein Titer Assay Kit, Acro Biosystems
Catalog # 103889-730
Supplier:  ACRO Biosystems
CAS Number:  
SARS-CoV-2 Nucleocapsid Protein Titer Assay Kit, Acro Biosystems
Catalog # 103889-730
Supplier:  ACRO Biosystems
Supplier Number:  RASA01096TESTS
CAS Number:  

Specifications

  • Description:
    SARS-CoV-2 Nucleocapsid Protein Titer Assay Kit
  • Size:
    96 tests
  • Cat. No.:
    103889-730
  • Storage temperature:
    –20 °C (avoid light)

Specifications

About this item

This kit is developed for detecting the SARS-CoV-2 Nucleocapsid Protein in cell supernatant or SARS-CoV-2 virus in vitro.

Nucleocapsid (N) protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein important for viral genome replication and modulation of cell signaling pathways. It was first identified by a research team while they were screening for ADP-ribosylated proteins during coronavirus (CoV) infection (Grunewald M. E., et al. 2017, Virology; 517: 62-68). The array of diverse functional activities accommodated in N protein makes it more than a structural protein but also an interesting target in the development of antiviral therapeutics. Because of the conservation of N protein sequence and its strong immunogenicity, N protein of coronavirus is chosen as a diagnostic tool.

Your experiment will include 5 simple steps:
a) Add your sample to the plate, take the SARS-CoV-2 Nucleocapsid protein as Control sample. The samples and Control sample are diluted by Lysis Buffer.
b) Add a diluted Secondary antibody biotin-anti-SARS-CoV-2 Nucleocapsid Antibody to the plate. The Secondary antody is diluted by Dilution Buffer.
c) Add a diluted Streptavidin-HRP to the plate.
d) Wash the plate and add TMB or other colorimetric HRP substrate.
e) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculate as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.

Certifications: ISO 9001 and ISO 13485.