Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SARS-CoV-2 Antibody IgG1 Titer Serologic Assay kit, Acro Biosystems
Anti-SARS-CoV-2 Antibody IgG1 Titer Serologic Assay kit, Acro Biosystems
Catalog # 103889-318
Supplier:  ACRO Biosystems
CAS Number:  
Anti-SARS-CoV-2 Antibody IgG1 Titer Serologic Assay kit, Acro Biosystems
Catalog # 103889-318
Supplier:  ACRO Biosystems
Supplier Number:  RAST01496TESTS
CAS Number:  

Specifications

  • Description:
    Anti-SARS-CoV-2 antibody IgG1 Titer serologic assay kit
  • Size:
    96 tests
  • Cat. No.:
    103889-318
  • Storage temperature:
    –20 °C (avoid light)

Specifications

About this item

This kit is developed for serologic test for IgG1 titer of Anti-SARS-CoV-2 antibody in serum/plasma in vitro.

The newly identified Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) poses continual threat to human health due to rapid transmission worldwide. Unprecedented vaccination campaigns have now begun with multiple candidates. To support these studies, ACRO has developed Anti-SARS-CoV-2 subtype antibody Serologic Assay kit including:IgG1, IgG2 IgG3, IgG4.

Your experiment will include four simple steps:
a) Add your sample to the plate, take the Anti-SARS-CoV-2 antibody as Control sample. The samples and Control sample are diluted by Dilution Buffer.
b) Add diluted Secondary antibody HRP-Mouse-Anti-Human IgG1 to the plate. The Secondary antibody is diluted by Dilution Buffer.
c) Wash the plate and add TMB or other colorimetric HRP substrate.
d) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculated as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.

Certifications: ISO 9001 and ISO 13485.