Order Entry
Puerto Rico
ContactUsLinkComponent
Monkey Anti-SARS-CoV-2 Antibody IgG Titer Serologic Assay Kit, Acro Biosystems
Monkey Anti-SARS-CoV-2 Antibody IgG Titer Serologic Assay Kit, Acro Biosystems
Catalog # 103889-148
Supplier:  ACRO Biosystems
CAS Number:  
Monkey Anti-SARS-CoV-2 Antibody IgG Titer Serologic Assay Kit, Acro Biosystems
Catalog # 103889-148
Supplier:  ACRO Biosystems
Supplier Number:  RAST01996TESTS
CAS Number:  

Specifications

  • Description:
    Monkey Anti-SARS-CoV-2 antibody IgG Titer serologic assay kit
  • Size:
    96 tests
  • Cat. No.:
    103889-148
  • Storage temperature:
    –20 °C (avoid light)

Specifications

About this item

This kit is developed for serologic test for Monkey IgG titer of Anti-SARS-CoV-2 antibody in serum/plasma in vitro.

The ongoing SARS-COV-2 pandemics has sickened 3.7 million people around the world, claiming over 1,000,000 lives. To deal with this global public health crisis, unprecedented efforts are being made to study the virus, SARS-CoV-2. At present, there are 193 new crown vaccine projects registered with the World Health Organization are being developed. To support these studies, ACRO has developed Monkey Anti-SARS-CoV-2 antibody IgG Serologic Assay kit.

Your experiment will include four simple steps:
a) Add your sample to the plate. The samples and Control sample are diluted by Dilution Buffer.
b) Add diluted Secondary antibody HRP-Anti-Goat anti monkey IgG to the plate. The Secondary antibody is diluted by Dilution Buffer.
c) Wash the plate and add TMB or other colorimetric HRP substrate.
d) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculate as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.

Certifications: ISO 9001 and ISO 13485.