Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SARS-CoV-2 Antibody IgG3 Titer Serologic Assay kit, Acro Biosystems
Anti-SARS-CoV-2 Antibody IgG3 Titer Serologic Assay kit, Acro Biosystems
Catalog # 103888-722
Supplier:  ACRO Biosystems
CAS Number:  
Anti-SARS-CoV-2 Antibody IgG3 Titer Serologic Assay kit, Acro Biosystems
Catalog # 103888-722
Supplier:  ACRO Biosystems
Supplier Number:  RAST01696TESTS
CAS Number:  

Specifications

  • Description:
    Anti-SARS-CoV-2 antibody IgG3 Titer serologic assay kit
  • Size:
    96 tests
  • Cat. No.:
    103888-722
  • Storage temperature:
    +2…+8 °C

Specifications

About this item

This kit is developed for serologic test for IgG3 titer of Anti-SARS-CoV-2 antibody in serum/plasma in vitro.

The ongoing SARS-COV-2 pandemics has sickened 3.7 million people around the world, claiming over 1000000 lives. To deal with this global public health crisis, unprecedented efforts are being made to study the virus, SARS-CoV-2. At present, there are 193 new crown vaccine projects registered with the World Health Organization are being developed. To support these studies, ACRO has developed Anti-SARS-CoV-2 subtype antibody Serologic Assay kit including:IgG1, IgG2 IgG3, IgG4.

Your experiment will include four simple steps:
a) Add your sample to the plate, take the Anti-SARS-CoV-2 antibody as Control sample. The samples and Control sample are diluted by Dilution Buffer.
b) Add diluted Secondary antibody HRP-Mouse-Anti-Human IgG3 to the plate. The Secondary antibody is diluted by Dilution Buffer.
c) Wash the plate and add TMB or other colorimetric HRP substrate.
d) Stop the substrate reaction by add diluted acid. Absorbance (OD) is calculated as the absorbance at 450 nm minus the absorbance at 650 nm to remove background prior to statistical analysis. The OD Value reflects the amount of antibody bound.

Certifications: ISO 9001 and ISO 13485.