Phenol Red Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
For the differentiation of microorganisms by carbohydrate fermentation.
- Contains a durham tube for gas detection
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.