Order Entry
Puerto Rico
ContactUsLinkComponent
Proteinase K Solution, Molecular Biology Grade, 20 mg/ml, MP Biomedicals
Proteinase K Solution, Molecular Biology Grade, 20 mg/ml, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
Proteinase K Solution, Molecular Biology Grade, 20 mg/ml, MP Biomedicals
Supplier:  MP Biomedicals

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    Yeast
  • Storage Conditions:
    2...8 °C
  • Reconstitution Instructions:
    Working concentrations of the enzyme range from 50 - 250 µg/ml
  • Enzyme Name:
    Proteinase K
  • Enzyme Activity:
    Specific activity >40 U/mg protein
  • Purity:
    >95%
  • Molecular Weight:
    29 kDa
  • Concentration:
    20 mg/ml
  • Formulation:
    Liquid in 10 mM Tris-HCl, 1 mM calcium acetate, pH 7.5 containing 50% glycerol
  • Grade:
    Molecular biology
  • Tested Applications:
    PCR, RNA purification, DNA purification, NGS, Sample preparation

Specifications

Related Information

Frequently Bought Together